Assay Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257009.1 | XP_021112668.1 | 3 - 4 | 351 | 98 | |
| XM_021257010.1 | XP_021112669.1 | 2 - 3 | 169 | 98 | ||
| XM_021257011.1 | XP_021112670.1 | 2 - 3 | 394 | 98 | ||
| XM_021257012.1 | XP_021112671.1 | 2 - 3 | 314 | 98 | ||
| XM_021257013.1 | XP_021112672.1 | 2 - 3 | 249 | 98 | ||
| XM_021257014.1 | XP_021112673.1 | 3 - 4 | 246 | 98 | ||
| XM_021257015.1 | XP_021112674.1 | 2 - 3 | 311 | 98 | ||
| XM_021257016.1 | XP_021112675.1 | 2 - 3 | 338 | 98 | ||
| XM_021257017.1 | XP_021112676.1 | 2 - 3 | 457 | 98 | ||

Fam192a
Genomic Map