Assay Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257009.1 | XP_021112668.1 | 6 - 7 | 703 | 64 | |
| XM_021257010.1 | XP_021112669.1 | 5 - 6 | 521 | 64 | ||
| XM_021257011.1 | XP_021112670.1 | 5 - 6 | 746 | 64 | ||
| XM_021257012.1 | XP_021112671.1 | 5 - 6 | 666 | 64 | ||
| XM_021257013.1 | XP_021112672.1 | 5 - 6 | 601 | 64 | ||
| XM_021257014.1 | XP_021112673.1 | 6 - 7 | 598 | 64 | ||
| XM_021257015.1 | XP_021112674.1 | 5 - 6 | 663 | 64 | ||
| XM_021257016.1 | XP_021112675.1 | 5 - 6 | 690 | 64 | ||
| XM_021257017.1 | XP_021112676.1 | 5 - 6 | 809 | 64 | ||
More Information
Set Membership: |
|

Fam192a
Genomic Map