Assay Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835721.3 | XP_004835778.1 | 4 - 5 | 708 | 148 | |
| XM_013075351.2 | XP_012930805.1 | 4 - 5 | 708 | 148 | ||
| XM_013075356.2 | XP_012930810.1 | 3 - 4 | 514 | 148 | ||
More Information
Set Membership: |
|

Gtpbp8
Genomic Map