Assay Details
| Gene Symbol: Psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872093.3 | XP_004872150.1 | 8 - 9 | 835 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psma4| Gene Symbol: Psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872093.3 | XP_004872150.1 | 8 - 9 | 835 | 71 | |
Set Membership: |
|