Assay Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834984.3 | XP_004835041.1 | 1 - 2 | 225 | 62 | |
| XM_004834985.3 | XP_004835042.1 | 1 - 2 | 226 | 62 | ||
| XM_021256882.1 | XP_021112541.1 | 1 - 2 | 228 | 62 | ||
| XM_021256886.1 | XP_021112545.1 | 1 - 2 | 228 | 62 | ||
| XM_021256900.1 | XP_021112559.1 | 1 - 2 | 228 | 62 | ||
More Information
Set Membership: |
|

Klhl18
Genomic Map