Assay Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077559.2 | XP_012933013.1 | 6 - 7 | 907 | 78 | |
| XM_021239726.1 | XP_021095385.1 | 5 - 6 | 564 | 78 | ||
| XM_021239727.1 | XP_021095386.1 | 6 - 7 | 814 | 78 | ||
| XM_021239728.1 | XP_021095387.1 | 6 - 7 | 937 | 78 | ||
More Information
Set Membership: |
|

Cfap126
Genomic Map