Assay Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867012.2 | XP_004867069.1 | 5 - 6 | 880 | 101 | |
| XM_004867013.2 | XP_004867070.1 | 4 - 5 | 685 | 101 | ||
| XM_021246258.1 | XP_021101917.1 | 6 - 7 | 1054 | 101 | ||
| XM_021246259.1 | XP_021101918.1 | 6 - 7 | 1032 | 101 | ||
| XM_021246260.1 | XP_021101919.1 | 6 - 7 | 1054 | 101 | ||
More Information
Set Membership: |
|

Tmem143
Genomic Map