Assay Details
| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834468.3 | XP_004834525.2 | 4 - 5 | 441 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004834468.3 transcripts.
Set Membership: |
|

Sucnr1
Genomic Map