Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866005.2 | XP_004866062.1 | 5 - 6 | 779 | 57 | |
XM_004866007.2 | XP_004866064.1 | 5 - 6 | 779 | 57 | ||
XM_013065923.2 | XP_012921377.1 | 4 - 5 | 659 | 57 | ||
XM_021244853.1 | XP_021100512.1 | 5 - 6 | 798 | 57 | ||
XM_021244854.1 | XP_021100513.1 | 5 - 6 | 800 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
