Assay Details
| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856992.3 | XP_004857049.1 | 2 - 3 | 416 | 70 | |
| XM_021238315.1 | XP_021093974.1 | 2 - 3 | 426 | 70 | ||
| XM_021238316.1 | XP_021093975.1 | 2 - 3 | 426 | 70 | ||
| XM_021238317.1 | XP_021093976.1 | 2 - 3 | 223 | 70 | ||
More Information
Set Membership: |
|

Unc119
Genomic Map