Assay Details
| Gene Symbol: Cd80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075206.2 | XP_012930660.2 | 2 - 3 | 670 | 60 | |
| XM_021263173.1 | XP_021118832.1 | 2 - 3 | 678 | 60 | ||
| XM_021263178.1 | XP_021118837.1 | 1 - 2 | 738 | 60 | ||
More Information
Set Membership: |
|

Cd80
Genomic Map