Assay Details
| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864075.2 | XP_004864132.1 | 3 - 4 | 1515 | 61 | |
| XM_021243861.1 | XP_021099520.1 | 3 - 4 | 1379 | 61 | ||
| XM_021243862.1 | XP_021099521.1 | 2 - 3 | 801 | 61 | ||
More Information
Set Membership: |
|

Nynrin
Genomic Map