Assay Details
| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844921.3 | XP_004844978.1 | 5 - 6 | 2097 | 70 | |
| XM_004844922.3 | XP_004844979.1 | 5 - 6 | 2087 | 70 | ||
| XM_021258451.1 | XP_021114110.1 | 5 - 6 | 2018 | 70 | ||
More Information
Set Membership: |
|

Usp44
Genomic Map