Assay Details
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013078313.2 | XP_012933767.1 | 2 - 3 | 637 | 78 | |
XM_021241384.1 | XP_021097043.1 | 3 - 4 | 626 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
