Assay Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843860.2 | XP_004843917.1 | 3 - 4 | 477 | 67 | |
| XM_021257529.1 | XP_021113188.1 | 3 - 4 | 392 | 67 | ||
| XM_021257530.1 | XP_021113189.1 | 2 - 3 | 501 | 67 | ||
More Information
Set Membership: |
|

Morn3
Genomic Map