Assay Details
| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870590.3 | XP_004870647.1 | 3 - 4 | 1368 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tefm| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870590.3 | XP_004870647.1 | 3 - 4 | 1368 | 97 | |
Set Membership: |
|