Assay Details
| Gene Symbol: Celf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855525.2 | XP_004855582.1 | 11 - 12 | 1321 | 87 | |
| XM_004855526.2 | XP_004855583.1 | 10 - 11 | 1258 | 87 | ||
| XM_013076125.2 | XP_012931579.1 | 10 - 11 | 1405 | 87 | ||
| XM_013076126.2 | XP_012931580.1 | 9 - 10 | 1231 | 87 | ||
| XM_013076127.2 | XP_012931581.1 | 9 - 10 | 1274 | 87 | ||
| XM_021236992.1 | XP_021092651.1 | 10 - 11 | 1405 | 87 | ||
| XM_021236993.1 | XP_021092652.1 | 10 - 11 | 1405 | 87 | ||
| XM_021236994.1 | XP_021092653.1 | 11 - 12 | 1318 | 87 | ||
| XM_021236995.1 | XP_021092654.1 | 10 - 11 | 1255 | 87 | ||
More Information
Set Membership: |
|

Celf6
Genomic Map