Assay Details
| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844601.3 | XP_004844658.1 | 2 - 3 | 286 | 70 | |
| XM_021258208.1 | XP_021113867.1 | 1 - 2 | 669 | 70 | ||
| XM_021258209.1 | XP_021113868.1 | 2 - 3 | 289 | 70 | ||
| XM_021258210.1 | XP_021113869.1 | 2 - 3 | 291 | 70 | ||
More Information
Set Membership: |
|

Tmem150a
Genomic Map