Assay Details
| Gene Symbol: Rrp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845463.3 | XP_004845520.2 | 4 - 5 | 485 | 97 | |
| XM_004845464.3 | XP_004845521.2 | 3 - 4 | 392 | 97 | ||
More Information
Set Membership: |
|

Rrp7a
Genomic Map