Assay Details
| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873931.3 | XP_004873988.1 | 9 - 10 | 1043 | 67 | |
| XM_013068939.2 | XP_012924393.1 | 9 - 10 | 1017 | 67 | ||
| XM_013068941.2 | XP_012924395.1 | 9 - 10 | 909 | 67 | ||
| XM_021251490.1 | XP_021107149.1 | 8 - 9 | 1029 | 67 | ||
More Information
Set Membership: |
|

Stoml2
Genomic Map