Assay Details
| Gene Symbol: Cd300a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860591.2 | XP_004860648.1 | 3 - 4 | 676 | 56 | |
| XM_004860592.2 | XP_004860649.1 | 3 - 4 | 676 | 56 | ||
More Information
Set Membership: |
|

Cd300a
Genomic Map