Assay Details
| Gene Symbol: Fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861529.2 | XP_004861586.1 | 2 - 3 | 327 | 70 | |
| XM_021241917.1 | XP_021097576.1 | 2 - 3 | 328 | 70 | ||
More Information
Set Membership: |
|

Fsip1
Genomic Map