Assay Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874664.2 | XP_004874721.1 | 5 - 6 | 1588 | 70 | |
| XM_021252077.1 | XP_021107736.1 | 5 - 6 | 2165 | 70 | ||
| XM_021252078.1 | XP_021107737.1 | 5 - 6 | 2166 | 70 | ||
More Information
Set Membership: |
|

Glis1
Genomic Map