Assay Details
| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875210.2 | XP_004875267.1 | 5 - 6 | 743 | 62 | |
| XM_013069460.2 | XP_012924914.1 | 6 - 7 | 1606 | 62 | ||
| XM_021252459.1 | XP_021108118.1 | 6 - 7 | 1460 | 62 | ||
| XM_021252460.1 | XP_021108119.1 | 7 - 8 | 1641 | 62 | ||
| XM_021252461.1 | XP_021108120.1 | 7 - 8 | 1836 | 62 | ||
| XM_021252462.1 | XP_021108121.1 | 6 - 7 | 1489 | 62 | ||
| XM_021252463.1 | XP_021108122.1 | 6 - 7 | 1424 | 62 | ||
| XM_021252464.1 | XP_021108123.1 | 6 - 7 | 1369 | 62 | ||
| XM_021252465.1 | XP_021108124.1 | 5 - 6 | 1330 | 62 | ||
| XM_021252466.1 | XP_021108125.1 | 4 - 5 | 454 | 62 | ||
| XM_021252467.1 | XP_021108126.1 | 4 - 5 | 381 | 62 | ||
More Information
Set Membership: |
|

March8
Genomic Map