Assay Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851597.3 | XP_004851654.1 | 3 - 4 | 512 | 72 | |
| XM_021263978.1 | XP_021119637.1 | 3 - 4 | 516 | 72 | ||
| XM_021263979.1 | XP_021119638.1 | 3 - 4 | 652 | 72 | ||
More Information
Set Membership: |
|

Dnajc24
Genomic Map