Assay Details
Gene Symbol: Gabpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854027.3 | XP_004854084.1 | 3 - 4 | 478 | 68 | |
XM_021265902.1 | XP_021121561.1 | 3 - 4 | 631 | 68 | ||
XM_021265903.1 | XP_021121562.1 | 3 - 4 | 483 | 68 | ||
XM_021265904.1 | XP_021121563.1 | 3 - 4 | 478 | 68 | ||
XM_021265905.1 | XP_021121564.1 | 3 - 4 | 478 | 68 | ||
XM_021265906.1 | XP_021121565.1 | 3 - 4 | 478 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
