Assay Details
| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868371.3 | XP_004868428.2 | 4 - 5 | 3062 | 61 | |
| XM_013066943.2 | XP_012922397.1 | 4 - 5 | 2951 | 61 | ||
| XM_013066944.2 | XP_012922398.1 | 5 - 6 | 3093 | 61 | ||
| XM_013066946.2 | XP_012922400.1 | 3 - 4 | 2739 | 61 | ||
| XM_021247425.1 | XP_021103084.1 | 6 - 7 | 3213 | 61 | ||
More Information
Set Membership: |
|

Sh3bp4
Genomic Map