Assay Details
| Gene Symbol: Yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249850.1 | XP_021105509.1 | 7 - 8 | 974 | 72 | |
| XM_021249851.1 | XP_021105510.1 | 7 - 8 | 1378 | 72 | ||
| XM_021249852.1 | XP_021105511.1 | 8 - 9 | 1115 | 72 | ||
| XM_021249853.1 | XP_021105512.1 | 7 - 8 | 1800 | 72 | ||
| XM_021249854.1 | XP_021105513.1 | 6 - 7 | 810 | 72 | ||
| XM_021249856.1 | XP_021105515.1 | 4 - 5 | 341 | 72 | ||
| XM_021249857.1 | XP_021105516.1 | 4 - 5 | 393 | 72 | ||
More Information
Set Membership: |
|

Yipf6
Genomic Map