Assay Details
Gene Symbol: Tmem167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853612.2 | XP_004853669.1 | 1 - 2 | 177 | 88 | |
XM_021265603.1 | XP_021121262.1 | 1 - 2 | 591 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853612.2, XM_021265603.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
