Assay Details
| Gene Symbol: Kcnj8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845770.3 | XP_004845827.1 | 2 - 3 | 750 | 90 | |
| XM_004845771.3 | XP_004845828.1 | 2 - 3 | 535 | 90 | ||
| XM_021259115.1 | XP_021114774.1 | 2 - 3 | 546 | 90 | ||
More Information
Set Membership: |
|

Kcnj8
Genomic Map