Assay Details
| Gene Symbol: Snrpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867140.3 | XP_004867197.1 | 1 - 2 | 187 | 75 | |
| XM_021245761.1 | XP_021101420.1 | 2 - 3 | 131 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004867140.3, XM_021245761.1 transcripts.
Set Membership: |
|

Snrpd2
Genomic Map