Assay Details
| Gene Symbol: Ly6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837863.3 | XP_004837920.3 | 1 - 2 | 112 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6d| Gene Symbol: Ly6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837863.3 | XP_004837920.3 | 1 - 2 | 112 | 75 | |