Assay Details
| Gene Symbol: Lamtor4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840217.2 | XP_004840274.1 | 3 - 4 | 338 | 58 | |
| XM_021254524.1 | XP_021110183.1 | 2 - 3 | 239 | 58 | ||
More Information
Set Membership: |
|

Lamtor4
Genomic Map