Assay Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836499.2 | XP_004836556.1 | 7 - 8 | 1169 | 66 | |
| XM_004836500.2 | XP_004836557.1 | 8 - 9 | 1516 | 66 | ||
| XM_004836501.2 | XP_004836558.1 | 6 - 7 | 1051 | 66 | ||
| XM_021240731.1 | XP_021096390.1 | 7 - 8 | 1357 | 66 | ||
More Information
Set Membership: |
|

Sgcd
Genomic Map