Assay Details
| Gene Symbol: Cldn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840143.3 | XP_004840200.1 | 4 - 5 | 1047 | 57 | |
| XM_021254474.1 | XP_021110133.1 | 5 - 6 | 1079 | 57 | ||
More Information
Set Membership: |
|

Cldn15
Genomic Map