Assay Details
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866126.3 | XP_004866183.1 | 9 - 10 | 1205 | 61 | |
XM_013066030.2 | XP_012921484.1 | 9 - 10 | 1325 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
