Assay Details
| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872904.3 | XP_004872961.2 | 1 - 2 | 334 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tctex1d4| Gene Symbol: Tctex1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872904.3 | XP_004872961.2 | 1 - 2 | 334 | 106 | |
Set Membership: |
|