Assay Details
| Gene Symbol: Lrfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852263.3 | XP_004852320.1 | 1 - 2 | 1784 | 93 | |
| XM_004852264.2 | XP_004852321.1 | 2 - 3 | 1765 | 93 | ||
More Information
Set Membership: |
|

Lrfn4
Genomic Map