Assay Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855369.3 | XP_004855426.1 | 1 - 2 | 199 | 62 | |
| XM_021236813.1 | XP_021092472.1 | 1 - 2 | 199 | 62 | ||
More Information
Set Membership: |
|

Star
Genomic Map