Assay Details
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842723.3 | XP_004842780.1 | 3 | 986 | 89 | |
XM_004842724.2 | XP_004842781.2 | 4 | 1128 | 89 | ||
XM_004842725.2 | XP_004842782.2 | 3 | 1138 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
