Assay Details
| Gene Symbol: Dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844144.2 | XP_004844201.1 | 1 - 2 | 885 | 91 | |
| XM_021257820.1 | XP_021113479.1 | 1 - 2 | 885 | 91 | ||
More Information
Set Membership: |
|

Dleu7
Genomic Map