Assay Details
| Gene Symbol: Znf425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860463.3 | XP_004860520.1 | 1 - 2 | 200 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004860463.3 transcripts.
Set Membership: |
|

Znf425
Genomic Map