Assay Details
| Gene Symbol: Znf425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860463.3 | XP_004860520.1 | 3 - 4 | 467 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf425| Gene Symbol: Znf425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860463.3 | XP_004860520.1 | 3 - 4 | 467 | 68 | |
Set Membership: |
|