Assay Details
| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846242.2 | XP_004846299.1 | 2 - 3 | 461 | 68 | |
| XM_021259596.1 | XP_021115255.1 | 2 - 3 | 428 | 68 | ||
More Information
Set Membership: |
|

Ddit4
Genomic Map