Assay Details
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072306.2 | XP_012927760.1 | 4 - 5 | 979 | 77 | |
XM_021258492.1 | XP_021114151.1 | 3 - 4 | 888 | 77 | ||
XM_021258493.1 | XP_021114152.1 | 4 - 5 | 1244 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
