Assay Details
| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263018.1 | XP_021118677.1 | 2 - 3 | 510 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldlrad2| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263018.1 | XP_021118677.1 | 2 - 3 | 510 | 71 | |
Set Membership: |
|