Assay Details
| Gene Symbol: Kiaa0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076868.2 | XP_012932322.1 | 17 - 18 | 6980 | 62 | |
| XM_013076869.2 | XP_012932323.1 | 10 - 11 | 1323 | 62 | ||
| XM_021238046.1 | XP_021093705.1 | 16 - 17 | 6873 | 62 | ||
| XM_021238047.1 | XP_021093706.1 | 9 - 10 | 1233 | 62 | ||
More Information
Set Membership: |
|

Kiaa0753
Genomic Map