Assay Details
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854780.3 | XP_004854837.1 | 3 - 4 | 1236 | 85 | |
| XM_021236307.1 | XP_021091966.1 | 5 - 6 | 1434 | 85 | ||
| XM_021236308.1 | XP_021091967.1 | 4 - 5 | 1323 | 85 | ||
More Information
Set Membership: |
|

Medag
Genomic Map