Assay Details
| Gene Symbol: Medag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854780.3 | XP_004854837.1 | 4 - 5 | 1518 | 102 | |
| XM_021236307.1 | XP_021091966.1 | 6 - 7 | 1716 | 102 | ||
| XM_021236308.1 | XP_021091967.1 | 5 - 6 | 1605 | 102 | ||
More Information
Set Membership: |
|

Medag
Genomic Map