Assay Details
Gene Symbol: Pin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872661.3 | XP_004872718.1 | 2 - 3 | 259 | 70 | |
XM_021250525.1 | XP_021106184.1 | 2 - 3 | 251 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
